missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Collagen I Native Protein

Product Code. 18667346 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18667346 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18667346 Supplier Novus Biologicals™ Supplier No. NBP262423
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Native Protein

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 mg/mL
For Use With (Application) ELISA
Formulation 0.5 M acetic acid
Gene ID (Entrez) 1277
Molecular Weight (g/mol) MolecularWeight-theroretical: 300 kDa
Quantity 0.5 mg
Storage Requirements Store at -80°C. Avoid freeze/thaw cycles.
Gene Alias Alpha-1 type I collagen, alpha1(I) procollagen, CAFYD, COL1A1, Collagen 1, collagen alpha 1 chain type I, collagen alpha-1(I) chain, collagen alpha-1(I) chain preproprotein, collagen of skin, tendon and bone, alpha-1 chain, collagen, type I, alpha 1, EDSARTH1, EDSC, OI1, OI2, OI3, OI4, pro-alpha-1 collagen type 1, type I proalpha 1, Type I Procollagen Alpha 1 Chain
Gene Symbol COL1A1
Research Category Cell Biology, Cellular Markers, Extracellular Matrix, Signal Transduction, Stem Cells
Purity or Quality Grade >90%, by SDS-PAGE
Protein Collagen I
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.