missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Collagen I Native Protein

Product Code. 18294824 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.01 g
Unit Size:
0.01g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18294824 0.01 g 0.01g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18294824 Supplier Novus Biologicals™ Supplier No. NBP2624270.01G
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: ELISA

Native rat collagen I

Source:Rat

Amino Acid Sequence:UniProt P02466, P02454

TRUSTED_SUSTAINABILITY

Specifications

Concentration LYOPH
For Use With (Application) ELISA
Formulation Lyophilised from aqueous 0.1 M acetic acid
Quantity 0.01 g
Storage Requirements Store at -80C. Avoid freeze-thaw cycles.
Reconstitution Reconstitute in ice cold 0.5 M acetic acid, pH 2.5. Solubility - 10 mg/mL.
Buffer Lyophilised from aqueous 0.1 M acetic acid
Purity or Quality Grade >90%, by SDS-PAGE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.