missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Collagen I/III Native Protein

Product Code. 18261743 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.01 g
Unit Size:
0.01g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18261743 0.01 g 0.01g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18261743 Supplier Novus Biologicals™ Supplier No. NBP2624280.01G
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: ELISA

Native mouse collagen I/III

Source:Murine tail tendons

Amino Acid Sequence:UniProt P08121, P11087, Q01149

TRUSTED_SUSTAINABILITY

Specifications

Concentration LYOPH
For Use With (Application) ELISA
Formulation Essentially salt free
Gene ID (Entrez) 1277
Molecular Weight (g/mol) 300 kDa
Quantity 0.01 g
Storage Requirements Store at 4°C. After reconstitution store at -20°C.
Gene Alias Alpha-1 type I collagen, Collagen 1/3, collagen alpha 1 chain type I, collagen alpha-1(I) chain, collagen of skin, tendon and bone, alpha-1 chain, collagen, type I, alpha 1, OI4, pro-alpha-1 collagen type 1
Gene Symbol COL1A1
Reconstitution Use 0.05-0.5 M acetic acid, pH 2.5 at 4°C. Dissolved collagen retains immunologic properties of native collagen types I and III. Thermal denaturation converts dissolved collagen to gelatin.
Buffer Essentially salt free
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.