missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Collagen I, bovine

Product Code. 10677174 Shop All Gibco Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mL
missing translation for 'unitSize':
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
10677174 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 10677174 missing translation for 'mfr' Gibco™ missing translation for 'supplierNo' A1064401

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Collagen I Bovine Protein

Collagen I, Bovine is a naturally occurring extra cellular matrix found in tendons and other connective tissues. It plays a key role in cell growth, differentiation, attachment, and migration. This solution is supplied at 5 mg⁄ml in a 10 ml vial providing the flexibility to dilute to lower concentrations. The detailed gelling procedure included in the package insert allows the user to form a firm, clear gel for 3D applications.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 5 mg/mL
For Use With (Application) Cell Culture
Quantity 10 mL
Product Type Collagen
Species Bovine
Content And Storage Store in refrigerator (2–8°C) and protect from light.
Endotoxin Level ≤10 EU/mL, ≤ 10 EU/ml
Shipping Condition Wet Ice
Protein Form Native
Form Liquid
Protein Subtype Collagen
Research Category Extracellular Matrix & Cell Adhesion
Product Line Gibco
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.