missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen I Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 195 publications
£451.00
Specifications
| Antigen | Collagen I alpha 1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, ELISA 1:5000-1:50000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:100, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:10-1:500, Immunocytochemistry 1:1600 |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
Collagen I Polyclonal specifically detects Collagen I in Human, Mouse, Rat, Porcine, Bacteria, Bovine, Rabbit samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, SDS-Page, Fluorophore-linked immunosorbent assay, Microarray.Specifications
| Collagen I alpha 1 | |
| Western Blot 1:1000, ELISA 1:5000-1:50000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:100, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:10-1:500, Immunocytochemistry 1:1600 | |
| Polyclonal | |
| Rabbit | |
| Cancer, Cell Cycle and Replication, Cytoskeleton Markers, Extracellular Matrix | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
| Alpha-1 type I collagen, Collagen 1, Collagen 1 alpha 1, collagen alpha 1 chain type I, collagen alpha-1(I) chain, collagen of skin, tendon and bone, alpha-1 chain, collagen, type I, alpha 1, Collagen1, Collagen-1, OI4, pro-alpha-1 collagen type 1 | |
| COL1A1 | |
| IgG | |
| Affinity Purified | |
| Typically less than 1% cross-reactivity against other types of collagens was detected by ELISA against purified standards. Some class-specific anti-collagens may be specific for three-dimensional epitopes which may result in diminished reactivity with denatured collagen or formalin-fixed, paraffin embedded tissues. This antibody reacts with most mammalian Type I collagens and has negligible cross-reactivity with Type II, III, IV, V or VI collagens. Non-specific cross-reaction of anti-collagen antibodies with other human serum proteins or non-collagen extracellular matrix proteins is negligible. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| P02452, P02453 | |
| 1277 | |
| Collagen Type I from human and bovine placenta by limited pepsin digestion and selective salt precipitation. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| 139 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title