missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen I Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-408H
This item is not returnable.
View return policy
Description
Collagen I Polyclonal antibody specifically detects Collagen I in Human, Mouse, Rat, Porcine, Bovine, Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
Specifications
| Collagen I | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| Collagen 1, Collagen I | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 1277 | |
| Human, Mouse, Rat, Pig, Bovine, Rabbit | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| HRP | |
| PBS with 0.05% Sodium Azide | |
| COL1A1 | |
| Collagen I from human and bovine placenta (Uniprot: P02452) | |
| 0.1 mL | |
| Primary | |
| Some class-specific anti-collagens may be specific for three-dimensional epitopes which may result in diminished reactivity with denatured collagen or formalin-fixed, paraffin embedded tissues. This antibody reacts with most mammalian Type I collagens and has expected cross-reactivity with Type III and negligible cross reactivity with Type II, IV, V or VI collagens. Non-specific cross-reaction of anti-collagen antibodies with other human serum proteins or non-collagen extracellular matrix proteins has not been tested. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction