missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen I alpha 1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92858-0.02ml
This item is not returnable.
View return policy
Description
Collagen I alpha 1 Polyclonal antibody specifically detects Collagen I alpha 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Collagen I alpha 1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Alpha-1 type I collagen, COL-IA1, Collagen 1, Collagen 1 alpha 1, collagen alpha 1 chain type I, collagen alpha-1(I) chain, collagen of skin, tendon and bone, alpha-1 chain, Collagen Type I Alpha 1 Chain, collagen, type I, alpha 1, Collagen1, Collagen-1, EDSARTH1, OI4, pro-alpha-1 collagen type 1, Type I Procollagen Alpha 1 Chain | |
| A synthetic peptide corresponding to a sequence within amino acids 1151-1250 of human Collagen I alpha 1 (NP_000079.2). PGKDGLNGLPGPIGPPGPRGRTGDAGPVGPPGPPGPPGPPGPPSAGFDFSFLPQPPQEKAHDGGRYYRADDANVVRDRDLEVDTTLKSLSQQIENIRSPEG | |
| 0.02 mL | |
| Cell Biology, Cellular Markers, Extracellular Matrix, Signal Transduction, Stem Cells | |
| 1277 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction