missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Colistin sulfate, MedChemExpress
Description
Colistin sulfate is a polypeptide antibiotic which inhibits gram-negative bacteria by binding to lipopolysaccharides and phospholipids in the outer cell membrane of gram-negative bacteria.
Specifications
Specifications
| Chemical Name or Material | Colistin sulfate |
| Physical Form | Solid |
| CAS | 1264-72-8 |
| Color | Off-White |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C52H98N16O13.2.5H2O4S |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | Polymyxin E sulfate |
| Solubility Information | H2O : 50 mg/mL (35.70 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?