missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ COLGALT2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5109571

Product Code. 17147503

  • £450.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: RFVAERDSED DGEEPVVFPE SPLQSPTVLV AVLARNAAHT LPHFLGCLER LDYPKSRMAI WAATDHNVDN TTEIFREWLK NVQRLYHYVE WRPMDEPESY PDEIGPKHWP TSRFAHVMKL RQAALRTARE KWSDYILFID VDNFLTNPQT LNLLIAENKT IVAPMLESRG LYSNFWCGIT PKGFYKRTPD YVQIREWKRT GCF.

GLT25D2 has a beta-galactosyltransferase activity and transfers beta-galactose to hydroxylysine residues on collagen.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

COLGALT2
Polyclonal
Unconjugated
COLGALT2
AI427652; C1orf17; ColGalT 2; Colgalt2; Collagen beta(1-O)galactosyltransferase 2; D730024P12; Glt25d2; glycosyltransferase 25 domain containing 2; glycosyltransferase 25 family member 2; Hydroxylysine galactosyltransferase 2; KIAA0584; Procollagen galactosyltransferase; procollagen galactosyltransferase 2; RGD1304702
Rabbit
Affinity Chromatography
RUO
23127, 269132
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
1.65 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Q6NVG7, Q8IYK4
COLGALT2
Recombinant fusion protein containing a sequence corresponding to amino acids 28-230 of human COLGALT2 (NP_055916.1).
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.