missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ COLEC12 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13415347
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13415347 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13415347 Supplier Invitrogen™ Supplier No. PA530835

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: Raji, K562. Predicted reactivity: Mouse (92%), Rat (91%), Chicken (83%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the C-lectin family, proteins that possess collagen-like sequences and carbohydrate recognition domains. This protein is a scavenger receptor, a cell surface glycoprotein that can bind to carbohydrate antigens on microorganisms facilitating their recognition and removal. In addition, these receptors can recognize oxidized phospholipids so they may also participate in removing oxidatively damaged or apoptotic cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COLEC12
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene Colec12
Gene Accession No. Q4V885, Q5KU26
Gene Alias CLP1; CL-P1; COLEC12; Collectin placenta protein 1; collectin subfamily member 12; collectin sub-family member 12; collectin-12; hCL-P1; NSR2; Nurse cell scavenger receptor 2; Scara4; Scavenger receptor class A member 4; scavenger receptor class A, member 4; scavenger receptor with C-type lectin; Srcl
Gene Symbols Colec12
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the Extracellular domain of human Colec12. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361289, 81035
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.