missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COLEC12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540550
This item is not returnable.
View return policy
Description
Peptide sequence: GLGGDGGGCG GGGSGGGGAP RREPVPFPSG SAGPGPRGPR ATESGKRMDC Sequence homology: Guinea Pig: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 93%.
This gene encodes a member of the C-lectin family, proteins that possess collagen-like sequences and carbohydrate recognition domains. This protein is a scavenger receptor, a cell surface glycoprotein that can bind to carbohydrate antigens on microorganisms facilitating their recognition and removal. In addition, these receptors can recognize oxidized phospholipids so they may also participate in removing oxidatively damaged or apoptotic cells.
Specifications
| COLEC12 | |
| Polyclonal | |
| Unconjugated | |
| Colec12 | |
| CLP1; CL-P1; COLEC12; Collectin placenta protein 1; collectin subfamily member 12; collectin sub-family member 12; collectin-12; hCL-P1; NSR2; Nurse cell scavenger receptor 2; Scara4; Scavenger receptor class A member 4; scavenger receptor class A, member 4; scavenger receptor with C-type lectin; Srcl | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 81035 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q5KU26 | |
| Colec12 | |
| Synthetic peptide directed towards the middle region of human COLEC12. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction