missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COLEC11 Rabbit anti-Human, Mouse, Rat, Zebrafish, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15269-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the collectin family of C-type lectins that possess collagen-like sequences and carbohydrate recognition domains. Collectins are secreted proteins that play important roles in the innate immune system by binding to carbohydrate antigens on microorganisms, facilitating their recognition and removal. The encoded protein binds to multiple sugars with a preference for fucose and mannose. Mutations in this gene are a cause of 3MC syndrome-2. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.Specifications
| COLEC11 | |
| Polyclonal | |
| Unconjugated | |
| COLEC11 | |
| CL K1, CL K1 I, CL K1 II, CL K1 IIa, CL K1 IIb, CLK1, COLEC11, Collectin 11, Collectin kidney protein 1, collectin sub family member 11 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 366588, 492459, 71693, 78989 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.42 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q3SXB8, Q5U3G1, Q9BWP8 | |
| Colec11 | |
| COLEC11 Fusion Protein Ag7374 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat, Zebrafish | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction