missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ Hose Barb Fitting Kit

Product Code. 11861080
Change view
Click to view available options
Diameter (Metric) Outer:
1.6 mm
Unit Size:
Pack of 240
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer unitSize
11861080 1.6 mm Pack of 240
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11861080 Supplier Masterflex™ Supplier No. 0636590

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Organize your workstation with convenient barbed fittings kits. Cole-Parmer™ Hose Barb Fitting Kits let you keep a variety of barbed fitting styles on hand. Polypropylene fittings are autoclavable. Include sturdy plastic case to keep fittings organized.

Contains the following part numbers and quantities:

06365-83; 1/16 in. ID Elbow, 10/Kit
06365-84; 3/32 in. ID Elbow, 10/Kit
06365-85; 1/8 in. ID Elbow, 10/Kit
06365-86; 3/32 in. ID Tee, 25/Kit
06365-91; 1/16 in. ID Straight Connector, 25/Kit
06365-92; 1/8 in. ID Straight Connector, 25/Kit
06365-93; 3/32 in. ID Straight Connector, 25/Kit
06365-94; 1/16 x 1/8 in. ID Reducer, 20/Kit
06365-95; 1/16 x 3/32 in. ID Reducer, 20/Kit
06365-96; 3/32 x 1/8 in. ID Reducer, 20/Kit
06365-97; 1/16 in. ID Tee, 25/Kit
06365-98; 1/8 in. ID Tee, 25/Kit

Specifications

Diameter (Metric) Outer 1.6 mm
Packaging Quantity 240 Pack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.