missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Cold Dissociation Enzyme Mix - Kidney

Product Code. 30057713 Shop All Cytiva Products
Click to view available options
:
Cell Culture Dissociation Reagent
This item is not returnable. View return policy

Product Code. 30057713

Brand: Cytiva 29733434

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A standalone kit, intended for the dissociation of kidney tissue in single-cell sequencing.

  • Works effectively at 4°C
  • Minimized cell death and stress (defined as: apoptosis pathway markers, macrophages, etc.) and improved transcriptional profile of the single cells for more accurate results in single-cell sequencing and analysis
  • Ready-made solution specifically for kidney tissue
  • Optimized for the dissociation of the different segments that are operative in kidney tissue
  • Optimized for use with the VIA Extractor™ tissue disaggregator
  • One of the few systems that can sustain a cold temperature in a controlled manner. Effective dissociation at cooler temperatures increases viability and minimizes cell aggregation
TRUSTED_SUSTAINABILITY

Specifications

Cell Line Enzyme, Kidney
Cell Type Enzyme, Kidney
Culture Type Enzyme, Kidney
Form Liquid
Product Type Cell Culture Dissociation Reagent
Reagent Type Enzyme
Sterility Non-sterile
With Additives None
Without Additives None
Formulation Liquid Buffer and Enzyme Solution
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

For research use only (RUO). Not for diagnostic use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.