missing translation for 'onlineSavingsMsg'
Learn More

Colchicine, MedChemExpress

Product Code. 16431816
Encompass_Preferred
Click to view available options
Quantity:
200 mg
500 mg
Unit Size:
200mg
500mg
This item is not returnable. View return policy

Product Code. 16431816

Brand: Medchem Express HY16569

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Colchicine is a tubulin inhibitor and a microtubule disrupting agent. Colchicine inhibits microtubule polymerization with an IC50 of 3 nM. Colchicine is also a competitive antagonist of the α3 glycine receptors (GlyRs).

Colchicine is a tubulin inhibitor and a microtubule disrupting agent. Colchicine inhibits microtubule polymerization with an IC50 of 3 nM. Colchicine is also a competitive antagonist of the α3 glycine receptors (GlyRs).

Specifications

Chemical Name or Material Colchicine
CAS 64-86-8
Color White
Purity Grade Notes Research
Molecular Formula C22H25NO6
Quantity 500 mg
Solubility Information DMSO : ≥ 48 mg/mL (120.17 mM) ∣H2O : ≥ 33.33 mg/mL (83.44 mM)
SMILES CC(N[C@H](C1=C2)CCC3=CC(OC)=C(OC)C(OC)=C3C1=CC=C(OC)C2=O)=O
Molecular Weight (g/mol) 399.44
Formula Weight 399.44
Percent Purity 98.62%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.