missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Colchicine, 97%

Product Code. 10174953 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
500mg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10174953 5 g Glass bottle 5g
10185230 500 mg Glass bottle 500mg
10388310 1 g Glass bottle 1g
13428488 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10174953 Supplier Thermo Scientific Chemicals Supplier No. 227120050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Colchicine, 7alphah-colchicine, CAS # 64-86-8, is a laboratory grade formulation of the anti-inflammatory plant alkaloid drug. | CAS: 64-86-8 | C22H25NO6 | 399.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Colchicine (7alphah-colchicine) is an alkaloid found in Colchicum autumnale that has been shown to have anti-gout and anti-inflammatory properties
  • Colchicine binds tubulin and prevents the migration of immune cells, reducing inflammation

Applications

  • Colchicine is an anti-mitotic; may be used in the study of cell proliferation and migration
  • Intended for in vivo and in vitro research applications
  • For research use only, not intended to treat disease
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-86-8
Molecular Formula C22H25NO6
Molecular Weight (g/mol) 399.44
MDL Number MFCD00078484,MFCD00078484
InChI Key IAKHMKGGTNLKSZ-INIZCTEOSA-N
Synonym colchicine, colchicina, colchicin, condylon, colchicinum, colchisol, colsaloid, colcin, colchineos, 7alphah-colchicine
PubChem CID 45038708
IUPAC Name 2,2,2-trideuterio-N-[(7S)-1,2,3-trimethoxy-9-oxo-10-(trideuteriomethoxy)-6,7-dihydro-5H-benzo[a]heptalen-7-yl]acetamide
SMILES COC1=C(OC)C(OC)=C2C(CC[C@H](NC(C)=O)C3=CC(=O)C(OC)=CC=C23)=C1

Specifications

Melting Point 150.0°C to 160.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. on anhydrous and solvent-free substance (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 14,IV,946
Merck Index 15,2455
Specific Rotation Condition − 250.00 (20.00°C c=1,alcohol)
Specific Rotation − 250.00
Solubility Information Solubility in water: 45g/l (20°C). Other solubilities: soluble in alcohol and chloroform,practically insoluble in petroleum ether,1g/220mL ether,1g/100mL benzene
Formula Weight 399.44
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Colchicine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Colchicine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Germ cell mutagenicity Category 1B
Hazard Statement
  • H340-May cause genetic defects.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.