missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COL9A3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559075
This item is not returnable.
View return policy
Description
Immunogen sequence: PGVPGITGKP GVPGKEASEQ RIRELCGGMI SEQIAQLAAH LRKPLAPGSI GRP Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
This gene encodes one of the three alpha chains of type IX collagen, the major collagen component of hyaline cartilage. Type IX collagen, a heterotrimeric molecule, is usually found in tissues containing type II collagen, a fibrillar collagen. Mutations in this gene are associated with multiple epiphyseal dysplasia type 3. [provided by RefSeq, Jan 2010].
Specifications
| COL9A3 | |
| Polyclonal | |
| Unconjugated | |
| COL9A3 | |
| alpha 3 type IX collagen; AV006866; cb367; CO9A3; COL9A3; Collagen alpha-3(IX) chain; collagen IX, alpha-3 polypeptide; collagen type IX alpha 3; collagen type IX alpha 3 chain; collagen type IX proteoglycan; collagen, type IX, alpha 3; DJ885L7.4.1; EDM3; IDD; MED; procollagen, type IX, alpha 3; sb:cb367; si:ch73-162j3.1; wu:fa04f01 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1299 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q14050 | |
| COL9A3 | |
| Recombinant protein corresponding to Human COL9A3. Recombinant protein control fragment (Product #RP-98343). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction