missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
COL9A1 Polyclonal antibody specifically detects COL9A1 in Human samples. It is validated for ELISA
Specifications
Specifications
| Applications | ELISA |
| Gene Alias | alpha-1 polypeptide, collagen, type IX, alpha 1 |
| Host Species | Rabbit |
| Purification Method | Antigen and protein A Affinity-purified |
| Quantity | 0.1 mL |
| Primary or Secondary | Primary |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?