missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ COL7A1 Monoclonal Antibody (C7)

Product Code. 17836322
Change view
Click to view available options
Quantity:
20 μL
200 μL
Unit Size:
200µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17836322 200 μL 200µL
17644336 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17836322 Supplier Invitrogen™ Supplier No. MA541570

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

COL7A1 Monoclonal Antibody for Western Blot

This gene encodes the alpha chain of type VII collagen. The type VII collagen fibril, composed of three identical alpha collagen chains, is restricted to the basement zone beneath stratified squamous epithelia. It functions as an anchoring fibril between the external epithelia and the underlying stroma. Mutations in this gene are associated with all forms of dystrophic epidermolysis bullosa. In the absence of mutations, however, an acquired form of this disease can result from an autoimmune response made to type VII collagen. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen COL7A1
Applications Western Blot
Classification Monoclonal
Clone C7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene COL7A1
Gene Accession No. Q02388
Gene Alias alpha-1 type VII collagen; AW209154; COL7A1; collagen alpha-1(VII) chain; collagen type VII alpha 1; collagen type VII alpha 1 chain; collagen vii; collagen VII, alpha-1 polypeptide; collagen, type VII, alpha 1; EBD1; EBDCT; EBR1; HGNC:2214; LC collagen; long chain collagen; long-chain collagen; NDNC8; procollagen, type VII, alpha 1
Gene Symbols COL7A1
Host Species Mouse
Immunogen Recombinant protein Collagen Type VII. The antigen corresponds to amino acid range 190-472 of the target protein.
Purification Method Protein A/G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1294
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.