missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COL4A6 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£359.00
Specifications
| Antigen | COL4A6 |
|---|---|
| Dilution | Western Blot 1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
COL4A6 Polyclonal specifically detects COL4A6 in Human samples. It is validated for Western Blot.Specifications
| COL4A6 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| collagen alpha 6 type IV, collagen alpha-6(IV) chain, collagen IV, alpha-6 polypeptide, collagen of basement membrane, alpha-6, collagen, type IV, alpha 6, CXDELq22.3, DELXq22.3, dJ889N15.4 (Collagen Alpha 6(IV)), EC 2.7.7.8, MGC88184 | |
| The immunogen is a synthetic peptide directed towards the middle region of human COL4A6 (NP_001838). Peptide sequence GIKGKSGLPGAPGFPGISGHPGKKGTRGKKGPPGSIVKKGLPGLKGLPGN | |
| Affinity purified |
| Western Blot 1.0 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS buffer, 2% sucrose | |
| 1288 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title