missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ COL4A3BP Antibody (1A1), Novus Biologicals™

Product Code. 18348409 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18348409 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18348409 Supplier Novus Biologicals™ Supplier No. H00010087M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

COL4A3BP Monoclonal antibody specifically detects COL4A3BP in Human samples. It is validated for ELISA,Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen COL4A3BP
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 1A1
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_112729
Gene Alias Ceramide transfer protein, CERTFLJ20597, CERTL, collagen type IV alpha-3-binding protein, collagen, type IV, alpha 3 (Goodpasture antigen) binding protein, Goodpasture antigen-binding protein, GPBPSTARD11ceramide transporter, hCERT, lipid-transfer protein CERTL, StARD11, StAR-related lipid transfer (START) domain containing 11, StAR-related lipid transfer protein 11, START domain containing 11, START domain-containing protein 11
Host Species Mouse
Immunogen COL4A3BP (NP_112729, 499 a.a. ∽ 598 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. TWIVCNFSVDHDSAPLNNRCVRAKINVAMICQTLVSPPEGNQEISRDNILCKITYVANVNPGGWAPASVLRAVAKREYPKFLKRFTSYVQEKTAGKPILF
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Immune System Diseases, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 10087
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.