missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COL4A3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144090
This item is not returnable.
View return policy
Description
COL4A3 Polyclonal Antibody for IHC (P), ELISA
Type IV collagen, the major structural component of basement membranes, is a multimeric protein composed of 3 alpha subunits. These subunits are encoded by 6 different genes, alpha 1 through alpha 6, each of which can form a triple helix structure with 2 other subunits to form type IV collagen. This gene encodes alpha 3. In the Goodpasture syndrome, autoantibodies bind to the collagen molecules in the basement membranes of alveoli and glomeruli. The epitopes that elicit these autoantibodies are localized largely to the non-collagenous C-terminal domain of the protein. A specific kinase phosphorylates amino acids in this same C-terminal region and the expression of this kinase is upregulated during pathogenesis. This gene is also linked to an autosomal recessive form of Alport syndrome. The mutations contributing to this syndrome are also located within the exons that encode this C-terminal region. Like the other members of the type IV collagen gene family, this gene is organized in a head-to-head conformation with another type IV collagen gene so that each gene pair shares a common promoter. [provided by RefSeq, Jun 2010].
Specifications
| COL4A3 | |
| Polyclonal | |
| Unconjugated | |
| COL4A3 | |
| COL4A3; collagen alpha-3; Collagen alpha-3(IV) chain; collagen IV, alpha-3 polypeptide; collagen type IV alpha 3 chain; collagen, type IV, alpha 3 (Goodpasture antigen); Goodpasture antigen; IV chain; Tumstatin | |
| Rabbit | |
| Protein G | |
| RUO | |
| 1285 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| Q01955 | |
| COL4A3 | |
| Recombinant Human Collagen alpha-3(IV) chain protein (1427-1668AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction