missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Col3a1, Rabbit anti-Rat, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova PAB13501.100uL
This item is not returnable.
View return policy
Specifications
| Col3a1 | |
| Polyclonal | |
| ELISA (>1:200) Immunohistochemistry (1:500) Immunofluorescence (1:80) The optimal working dilution should be determined by the end user. | |
| collagen, type III, alpha 1 | |
| Col3a1 | |
| Native purified Col3a1 from rat skin. | |
| RUO | |
| 84032 | |
| Mouse, Rat | |
| Antibody |
| ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Radioimmune Assays (RIA) | |
| Unconjugated | |
| In phosphate buffer | |
| MGC93704 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| This antibody reacts with rat collagen type III 100%, rat collagen I <0.1%; rat collagen type V <10%; rat elastin <0.1% (solid phase RIA at 1:50 dilution). | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction