missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COL28A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583683
This item is not returnable.
View return policy
Description
Immunogen sequence: SESLSVTRDQ DEDDKAPEPT WADDLPATTS SEATTTPRPL LSTPVDGAED PRCLEALKPG NCGEYVVRWY YDKQVNSCAR FWFSGCNGSG NRFNSEKECQ ET Highest antigen sequence indentity to the following orthologs: Mouse - 57%, Rat - 58%.
COL28A1 belongs to a class of collagens containing von Willebrand factor (VWF; MIM 613160) type A (VWFA) domains (Veit et al., 2006 [PubMed 16330543]).[supplied by OMIM, Nov 2010].
Specifications
| COL28A1 | |
| Polyclonal | |
| Unconjugated | |
| COL28A1 | |
| COL28; COL28A1; Collagen alpha-1(XXVIII) chain; collagen type XXVIII alpha 1 chain; collagen, type XXVIII, alpha 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 340267 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q2UY09 | |
| COL28A1 | |
| Recombinant protein corresponding to Human COL28A1. Recombinant protein control fragment (Product #RP-100347). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction