missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COL26A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA537248
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 45 and 55 kDa. Purity is >95% by SDS-PAGE.
COL26A1 (EMID2) is a 441 amino acid protein encoded by a gene mapping to human chromosome 7. Chromosome 7 is approximately 158 milllion bases long, encodes over 1,000 genes and makes up about 5% of the human genome. Chromosome 7 has been linked to osteogenesis imperfecta, Pendred syndrome, lissencephaly, citrullinemia and Shwachman-Diamond syndrome. The deletion of a portion of the q arm of chromosome 7 is associated with Williams-Beuren syndrome, a condition characterized by mild mental retardation, an unusual comfort and friendliness with strangers, and an elfin appearance. Deletions of portions of the q arm of chromosome 7 are also seen in a number of myeloid disorders including cases of acute myelogenous leukemia and myelodysplasia.
Specifications
| COL26A1 | |
| Polyclonal | |
| Unconjugated | |
| COL26A1 | |
| 9430032K24Rik; alpha-1 type XXVI collagen; BC002218; Col26a; COL26A1; Collagen alpha-1(XXVI) chain; collagen type XXVI alpha 1; Collagen XXVI; collagen, type XXVI, alpha 1; EMI domain containing 2; EMI domain-containing protein 2; EMI6; EMID2; emilin and multimerin domain-containing protein 2; EMU2; procollagen, type XXVI, alpha 1; SH2B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 136227, 685612 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96A83 | |
| COL26A1 | |
| A synthetic peptide corresponding to residues in human COL26A1. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction