missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COL22A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570815
This item is not returnable.
View return policy
Description
COL22A1 Polyclonal Antibody for Western Blot
This gene encodes member of the collagen family which is thought to contribute to the stabilization of myotendinous junctions and strengthen skeletal muscle attachments during contractile activity. It belongs to the fibril-associated collagens with interrupted triple helix (FACIT) subset of the collagen superfamily, which associate with collagen fibers through their C-terminal collagenous domains and mediate protein-protein interactions through their N-terminal noncollagenous domains. The encoded protein is deposited in the basement membrane zone of the myotendinous junction which is present only at the tissue junctions of muscles, tendons, the heart, articular cartilage, and skin. A knockdown of the orthologous zebrafish gene induces a muscular dystrophy by disruption of the myotendinous junction.
Specifications
| COL22A1 | |
| Polyclonal | |
| Unconjugated | |
| COL22A1 | |
| 2310067L16Rik; C80743; COL22A1; Collagen alpha-1(XXII) chain; collagen type XXII alpha 1; collagen type XXII alpha 1 chain; collagen, type XXII, alpha 1; procollagen, type XXII, alpha 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 169044 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8NFW1 | |
| COL22A1 | |
| synthetic peptide directed towards the N-terminal region of human COL22A1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction