missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COL21A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556675
This item is not returnable.
View return policy
Description
Immunogen sequence: AKSSRFLTKI AVVLTDGKSQ DDVKDAAQAA RDSKITLFAI GVGSETEDAE LRAIANKPSS TYVFYVEDYI AISKIREVMK Highest antigen sequence identity to the following orthologs: Mouse - 45%, Rat - 45%.
COL17A1 is the alpha chain of type XVII collagen. Unlike most collagens, collagen XVII is a transmembrane protein, and a structural component of hemidesmosomes, multiprotein complexes at the dermal-epidermal basement membrane zone that mediate adhesion of keratinocytes to the underlying membrane. COL17A1 is a member of the FACIT collagen family (fibril-associated collagens with interrupted helices). Moreover, COL17A1 is localized to tissues containing type I collagen so, like other members of this collagen family, it may serve to maintain the integrity of the extracellular matrix. Diseases associated with COL17A1 dysfunction include epithelial recurrent erosion dystrophy and epidermolysis bullosa.
Specifications
| COL21A1 | |
| Polyclonal | |
| Unconjugated | |
| COL21A1 | |
| alpha 1 chain-like collagen; COL1AL; COL21A1; COLA1L; Collagen alpha-1(XXI) chain; collagen type XXI alpha 1 chain; collagen, type XXI, alpha 1; FP633 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 81578 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96P44 | |
| COL21A1 | |
| Recombinant protein corresponding to Human COL21A1. Recombinant protein control fragment (Product #RP-94805). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction