missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COL20A1 Antibody (1F5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00057642-M01
This item is not returnable.
View return policy
Description
COL20A1 Monoclonal antibody specifically detects COL20A1 in Human samples. It is validated for ELISA, Sandwich ELISA
Specifications
| COL20A1 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| collagen alpha-1(XX) chain, collagen, type XX, alpha 1, collagen-like protein, KIAA1510bA261N11.4 | |
| KIAA1510 (AAH19637, 1 a.a. ∽ 155 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MRGLEGTAGLPGPPGPRGFQGMAGARGTSGERGPPGTVGPTGLPGPKGERGEKGEPQSLATLYQLVSQACESAIQTHVSKFDSFHENTRPPMPILEQKLEPGTEPLGSPGTRSKALVPGEWGRGGRHLEGRGEPGAVGQMGSPGQQGASTQGLWE | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| ELISA, Sandwich ELISA | |
| 1F5 | |
| ELISA, Sandwich ELISA | |
| AAH19637 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 57642 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction