missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COL1A2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596426
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Positive Samples: Mouse skin, Mouse skeletal muscle, Rat skin; Cellular Location: extracellular matrix, extracellular space Immunogen sequence: TGDPGKNGDK GHAGLAGARG APGPDGNNGA QGPPGPQGVQ GGKGEQGPPG PPGFQGLPGP SGPAGEVGKP GERGLHGEFG LPGPAGPRGE RGPPGESGAA.
This gene encodes the pro-alpha2 chain of type I collagen whose triple helix comprises two alpha1 chains and one alpha2 chain. Type I is a fibril-forming collagen found in most connective tissues and is abundant in bone, cornea, dermis and tendon. Mutations in this gene are associated with osteogenesis imperfecta types I-IV, Ehlers-Danlos syndrome type VIIB, recessive Ehlers-Danlos syndrome Classical type, idiopathic osteoporosis, and atypical Marfan syndrome. Symptoms associated with mutations in this gene, however, tend to be less severe than mutations in the gene for the alpha1 chain of type I collagen (COL1A1) reflecting the different role of alpha2 chains in matrix integrity. Three transcripts, resulting from the use of alternate polyadenylation signals, have been identified for this gene.
Spécification
| COL1A2 | |
| Polyclonal | |
| Unconjugated | |
| COL1A2 | |
| AA960264; AI325291; alpha 2 type I procollagen; alpha 2(I) collagen; alpha 2(I) procollagen; alpha 2(I)-collagen; alpha2 chain of type I collagen; alpha-2 type I collagen; alpha-2(1) collagen; COL1; COL1A2; Col1a-2; COLA; Cola2; Cola-2; collagen; collagen a2; collagen alpha 2(I) chain precursor; collagen alpha-2; collagen alpha-2 chain precursor; collagen alpha-2(I) chain; collagen COL1A2; Collagen I; collagen I, alpha-2 polypeptide; collagen of skin, tendon and bone, alpha-2 chain; collagen type I alpha 2 chain; collagen, type I, alpha 2; OI4; oim; osteogenesis imperfecta; precollagen alpha-2; procollagen, type I, alpha 2; type I collagen alpha-2 chain; type I procollagen | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 1278, 12843, 84352 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1.05 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P02466, P08123, Q01149 | |
| COL1A2 | |
| Synthetic peptide | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu