missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ COL16A1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17239403 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17239403 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17239403 Supplier Invitrogen™ Supplier No. PA5116903

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Samples: PC-3, Mouse kidney, Mouse heart Immunogen sequence: PRAEEARGDN SEGDPGCVGS PGLPGPPGLP GQRGEEGPPG MRGSPGPPGP IGPPGFPGAV GSPGLPGLQG ERGLTGLTGD KGEPGPPGQP GYPGATGPPG L.

This gene encodes the alpha chain of type XVI collagen, a member of the FACIT collagen family (fibril-associated collagens with interrupted helices). Members of this collagen family are found in association with fibril-forming collagens such as type I and II, and serve to maintain the integrity of the extracellular matrix. High levels of type XVI collagen have been found in fibroblasts and keratinocytes, and in smooth muscle and amnion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COL16A1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.19 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene COL16A1
Gene Accession No. Q07092, Q8BLX7
Gene Alias [a]1 (XVI) collagen; 2700007F12Rik; 447AA; A530052M23Rik; AI838662; alpha 1 type XVI collagen; CA1F; col16a1; Collagen alpha-1(XVI) chain; collagen type XVI alpha 1 chain; collagen XVI, alpha-1 polypeptide; collagen, type XVI, alpha 1; FP1572; procollagen type XVI alpha 1; procollagen, type XVI, alpha 1
Gene Symbols COL16A1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1000-1100 of human COL16A1 (NP_0018473).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 107581, 1307, 366474
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.