missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COL15A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115179
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Endostatin.
This gene encodes the alpha chain of type XV collagen, a member of the FACIT collagen family (fibril-associated collagens with interrupted helices). Type XV collagen has a wide tissue distribution but the strongest expression is localized to basement membrane zones so it may function to adhere basement membranes to underlying connective tissue stroma. The proteolytically produced C-terminal fragment of type XV collagen is restin, a potentially antiangiogenic protein that is closely related to endostatin. Mouse studies have shown that collagen XV deficiency is associated with muscle and microvessel deterioration.
Specifications
| COL15A1 | |
| Polyclonal | |
| Unconjugated | |
| COL15A1 | |
| COFA1; COL15A1; Collagen alpha-1(XV) chain; collagen type XV alpha 1 chain; collagen type XV proteoglycan; collagen XV, alpha-1 polypeptide; collagen, type XV, alpha 1; endostatin-XV; procollagen type XV; procollagen, type XV; Related to endostatin; Restin; Restin-2; Restin-3; Restin-4; Restin-I; Restin-II; Restin-III; Restin-IV; type XV collagen | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12819, 1306, 298069 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| O35206, P39059 | |
| COL15A1 | |
| A synthesized peptide derived from human COL15A1(Accession P39059), corresponding to amino acid residues A272-I322. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction