missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COL12A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552655
This item is not returnable.
View return policy
Description
Immunogen sequence: VTTPPNQRRR TLENLIPDTK YEVSVIPEYF SGPGTPLTGN AATEEVRGNP RDLRVSDPTT STMKLSWSGA PGKVKQYLVT YTPVAGGETQ EVTVRGDTTN TVLQGLKEGT QYALSVTALY ASGAG Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.
This gene encodes the alpha chain of type XII collagen, a member of the FACIT (fibril-associated collagens with interrupted triple helices) collagen family. Type XII collagen is a homotrimer found in association with type I collagen, an association that is thought to modify the interactions between collagen I fibrils and the surrounding matrix. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq, Jul 2008].
Specifications
| COL12A1 | |
| Polyclonal | |
| Unconjugated | |
| COL12A1 | |
| BA209D8.1; BTHLM2; COL12A1; COL12A1L; collagen alpha-1(XII) chain; collagen type XII alpha 1 chain; collagen type XII proteoglycan; collagen, type XII, alpha 1; DJ234P15.1; UCMD2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1303 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q99715 | |
| COL12A1 | |
| Recombinant protein corresponding to Human COL12A1. Recombinant protein control fragment (Product #RP-89835). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction