missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coilin Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-17696-100UL
This item is not returnable.
View return policy
Description
Coilin Polyclonal antibody specifically detects Coilin in Human samples. It is validated for Western Blot
Specifications
| Coilin | |
| Polyclonal | |
| Western Blot 0.04-0.4 ug/ml | |
| CLN80p80-coilin, coilin, coilin p80, p80 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: ASETVRLRLQFDYPPPATPHCTAFWLLVDLNRCRVVTDLISLIRQRFGFSSGAFLGLYLEGGLLPPAESARLVRDNDCLRVKLEERGVAENSVVISNGDINLSLRKAKKRAFQLEEGEETEPDC | |
| 100 μg | |
| Neuroscience | |
| 8161 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction