missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COH29, MedChemExpress
Description
COH29 (RNR Inhibitor COH29) is a potent ribonucleotide reductase (RNR) inhibitor with anticancer activity. COH29 inhibits α and β subunit of RNR with IC50s of 16 μM.
Specifications
Specifications
| Chemical Name or Material | COH29 |
| CAS | 1190932-38-7 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C22H16N2O5S |
| Quantity | 10 mg |
| Synonym | RNR Inhibitor COH29 |
| Solubility Information | DMSO : 50 mg/mL (118.92 mM; Need ultrasonic) |
| SMILES | O=C(NC1=NC(C2=CC=C(O)C(O)=C2)=C(C3=CC=CC=C3)S1)C4=CC=C(O)C(O)=C4 |
| Molecular Weight (g/mol) | 420.44 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?