missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COG6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35123-100ul
This item is not returnable.
View return policy
Description
COG6 Polyclonal antibody specifically detects COG6 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| COG6 | |
| Polyclonal | |
| Western Blot 1:1000 - 1:2000, ELISA | |
| COG complex subunit 6, component of oligomeric golgi complex 6KIAA1134COD2complexed with Dor1p 2, conserved oligomeric Golgi complex protein 6, conserved oligomeric Golgi complex subunit 6, DKFZp313D191 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 508-657 of human COG6 (NP_065802.1).,, Sequence:, MKTTLALFEFTDRRLEMLQFQIEAHLDTLINEQASYVLTRVGLSYIYNTVQQHKPEQGSLANMPNLDSVTLKAAMVQFDRYLSAPDNLLIPQLNFLLSATVKEQIVKQSTELVCRAYGEVYAAVMNPINEYKDPENILHRSPQQVQTLLS | |
| 100 μL | |
| Golgi Apparatus Markers | |
| 57511 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction