missing translation for 'onlineSavingsMsg'
Learn More

COG6 Antibody, Novus Biologicals™

Product Code. 18392915 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18392915 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18392915 Supplier Novus Biologicals Supplier No. NBP318428

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

COG6 Polyclonal antibody specifically detects COG6 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen COG6
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:2000 - 1:10000, Immunoprecipitation 20 ug/mg lysate
Formulation Tris-citrate/phosphate pH 7 -8.
Gene Alias COG complex subunit 6, component of oligomeric golgi complex 6KIAA1134COD2complexed with Dor1p 2, conserved oligomeric Golgi complex protein 6, conserved oligomeric Golgi complex subunit 6, DKFZp313D191
Host Species Rabbit
Immunogen Recognizes region between residues Between Between 607 and 657..
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Golgi Apparatus Markers
Primary or Secondary Primary
Gene ID (Entrez) 57511
Target Species Human, Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.