missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COG5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£380.00
Specifications
| Antigen | COG5 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18264299
|
Novus Biologicals
NBP1-91374 |
100 μL | |||||||
Description
COG5 Polyclonal specifically detects COG5 in Human samples. It is validated for Western Blot.Specifications
| COG5 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| component of oligomeric golgi complex 5, FLJ44289 | |
| COG5 | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RUO | |
| NP_006339 | |
| 10466 | |
| Synthetic peptide directed towards the N terminal of human COG5. Peptide sequence MGWVGGRRRDSASPPGRSRSAADDINPAPANMEGGGGSVAVAGLGARGSG. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title