missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COG4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£330.00 - £486.00
Specifications
| Antigen | COG4 |
|---|---|
| Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18468861
|
Novus Biologicals
NBP2-30718-25ul |
25 μL |
£330.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18056374
|
Novus Biologicals
NBP2-30718 |
0.1 mL |
£486.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
COG4 Polyclonal specifically detects COG4 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| COG4 | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| Q9H9E3 | |
| 25839 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: ILDLKFCMDGVQTALRSEDYEQAAAHIHRYLCLDKSVIELSRQGKEGSMIDANLKLLQEAEQRLKAIVAEKFAIATKEGDLPQVERFFKIFPLL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Golgi Apparatus Markers | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| CDG2J, COD1complexed with Dor1p, COG complex subunit 4, component of oligomeric golgi complex 4DKFZp586E1519, conserved oligomeric Golgi complex protein 4, conserved oligomeric Golgi complex subunit 4, DKFZP586E1519 | |
| COG4 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title