missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COG3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11130-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a component of the conserved oligomeric Golgi (COG) complex which is composed of eight different subunits and is required for normal Golgi morphology and localization. Defects in the COG complex result in multiple deficiencies in protein glycosylation. The protein encoded by this gene is involved in ER-Golgi transport.Specifications
| COG3 | |
| Polyclonal | |
| Unconjugated | |
| COG3 | |
| COG complex subunit 3, COG3, p94, SEC34 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 338337, 361073, 83548 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8CI04, Q96JB2 | |
| Cog3 | |
| COG3 Fusion Protein Ag1610 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction