missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COG3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584200
This item is not returnable.
View return policy
Description
Immunogen sequence: DGQLFLIKHL LILREQIAPF HTEFTIKEIS LDLKKTRDAA FKILNPMTVP RFFRLNSNNA LIEFLLEGTP EIREHYLDSK KDVDRHLKSA CEQFIQQQT Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.
This gene encodes a component of the conserved oligomeric Golgi (COG) complex which is composed of eight different subunits and is required for normal Golgi morphology and localization. Defects in the COG complex result in multiple deficiencies in protein glycosylation. The protein encoded by this gene is involved in ER-Golgi transport.
Specifications
| COG3 | |
| Polyclonal | |
| Unconjugated | |
| COG3 | |
| COG complex subunit 3; COG3; component of oligomeric golgi complex 2; component of oligomeric golgi complex 3; Conserved oligomeric Golgi complex subunit 3; E430004N23; p94; SEC34; tethering factor SEC34; vesicle-docking protein SEC34 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 83548 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96JB2 | |
| COG3 | |
| Recombinant protein corresponding to Human COG3. Recombinant protein control fragment (Product #RP-108403). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction