missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COG3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-36540
This item is not returnable.
View return policy
Description
COG3 Polyclonal specifically detects COG3 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| COG3 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| COG complex subunit 3, component of oligomeric golgi complex 3p94, SEC34conserved oligomeric Golgi complex subunit 3, tethering factor SEC34, Vesicle-docking protein SEC34 homolog | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 83548 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg | |
| EAX08741.1 | |
| COG3 | |
| The immunogen recognized by the antibody maps to a region between residue 1 to 50 of human Component of Oligomeric Golgi Complex 3 using the numbering given in entry EAX08741.1. | |
| 1 mL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction