missing translation for 'onlineSavingsMsg'
Learn More

COG2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£145.00 - £417.00

Specifications

Antigen COG2
Dilution Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:100
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30232058
View Documents
Novus Biologicals
NBP3-38235-100ul
100 μL
£417.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30229596
View Documents
Novus Biologicals
NBP3-38235-20ul
20 μL
£145.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

COG2 Polyclonal antibody specifically detects COG2 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications

Specifications

COG2
ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Golgi Apparatus Markers
PBS (pH 7.3), 50% glycerol
22796
IgG
Affinity purified
Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:100
Polyclonal
Purified
RUO
Human, Mouse, Rat
component of oligomeric golgi complex 2COG complex subunit 2, conserved oligomeric Golgi complex protein 2, conserved oligomeric Golgi complex subunit 2, LDLCbrefeldin A-sensitive, peripheral Golgi protein, low density lipoprotein receptor defect C complementing, Low density lipoprotein receptor defect C-complementing protein
Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human COG2 (NP_031383.1).,, Sequence:, MEKSRMNLPKGPDTLCFDKDEFMKEDFDVDHFVSDCRKRVQLEELRDDLELYYKLLKTAMVELINKDYADFVNLSTNLVGMDKALNQLSVPLGQLREEVLSLRSSVSEGIRAVDERMSKQEDIRKKKMCVLRLIQVIRSVEKIEKILNSQSSKETSALEASSPLLTGQILERIATEFNQLQFHAVQSKGMPLLDKVRPRIAGITAMLQQSLEGLLLEGLQTSDVDIIRHCLRTYATIDKTRDAEALVGQVLVKPYIDEVIIEQFVESHPNGLQVMYNKLL
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Filmer
SDS
Dokument

Dokument

Produktspecifikationer
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.