missing translation for 'onlineSavingsMsg'
Learn More

COG2 Antibody (4C8), Novus Biologicals™

Product Code. 18389329 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18389329 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18389329 Supplier Novus Biologicals Supplier No. H00022796M09

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

COG2 Monoclonal antibody specifically detects COG2 in Human samples. It is validated for Western Blot, ELISA, KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen COG2
Applications Western Blot, ELISA, KnockDown
Classification Monoclonal
Clone 4C8
Conjugate Unconjugated
Dilution Western Blot 1:500RNAi Validation, ELISA, Knockdown Validated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_031383
Gene Alias component of oligomeric golgi complex 2COG complex subunit 2, conserved oligomeric Golgi complex protein 2, conserved oligomeric Golgi complex subunit 2, LDLCbrefeldin A-sensitive, peripheral Golgi protein, low density lipoprotein receptor defect C complementing, Low density lipoprotein receptor defect C-complementing protein
Host Species Mouse
Immunogen COG2 (NP_031383, 639 a.a. ~ 738 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. LSESTHKYYETVSDVLNSVKKMEESLKRLKQARKTTPANPVGPSGGMSDDDKIRLQLALDVEYLGEQIQKLGLQASDIKSFSALAELVAAAKDQATAEQP
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Golgi Apparatus Markers
Primary or Secondary Primary
Gene ID (Entrez) 22796
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.