missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COG1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556085
This item is not returnable.
View return policy
Description
Immunogen sequence: FCSALDSKLK VKLDDLLAYL PSDDSSLPKD VSPTQAKSSA FDRYADAGTV QEMLRTQSVA CIKHIVDCIR AELQSIEEGV QGQQDALNSA KLHSVLFMA Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 76%.
The protein encoded by this gene is one of eight proteins which form a Golgi-localized complex required for normal Golgi morphology and function. It is thought that this protein is required for steps in the normal medial and trans Golgi-associated processing of glycoconjugates and plays a role in the organization of the Golgi-localized complex.
Specifications
| COG1 | |
| Polyclonal | |
| Unconjugated | |
| COG1 | |
| 6030445I15; CDG2G; COG complex subunit 1; Cog1; Component of oligomeric Golgi complex 1; conserved oligomeric Golgi complex protein 1; conserved oligomeric Golgi complex subunit 1; DKFZp762L1710; KIAA1381; Ldlb; low density lipoprotein B; low density lipoprotein receptor defect B complementing; low density lipoprotein receptor defect B-complementing protein; mKIAA1381 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9382 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WTW3 | |
| COG1 | |
| Recombinant protein corresponding to Human COG1. Recombinant protein control fragment (Product #RP-93371). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction