Learn More
COG1 Mouse, anti-Human, Clone: 2D8D11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68642-1-IG-20UL
This item is not returnable.
View return policy
Description
The COG1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human COG1. This antibody recognizes human, rabbit antigen. The COG1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| COG1 | |
| 2D8D11 | |
| CDG2G, COG complex subunit 1, COG1, DKFZp762L1710, KIAA1381, LDLB | |
| 20 μL | |
| 9382 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rabbit | |
| 150 kDa | |
| IgG1 |