missing translation for 'onlineSavingsMsg'
Learn More

COG1 Antibody, Novus Biologicals™

Product Code. 18378528 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18378528 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18378528 Supplier Novus Biologicals Supplier No. H00009382B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

COG1 Polyclonal antibody specifically detects COG1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen COG1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation PBS (pH 7.4)
Gene Accession No. NP_061184.1
Gene Alias COG complex subunit 1, component of oligomeric golgi complex 1CDG2G, conserved oligomeric Golgi complex subunit 1, KIAA1381DKFZp762L1710, LDLBconserved oligomeric Golgi complex protein 1, low density lipoprotein receptor defect B complementing
Host Species Mouse
Immunogen COG1 (NP_061184.1, 1 a.a. - 980 a.a.) full-length human protein. MATAATSPALKRLDLRDPAALFETHGAEEIRGLERQVRAEIEHKKEELRQMVGERYRDLIEAADTIGQMRRCAVGLVDAVKATDQYCARLRQAGSAAPRPPRAQQPQQPSQEKFYSMAAQIKLLLEIPEKIWSSMEASQCLHATQLYLLCCHLHSLLQLDSSSSRYSPVLSRFPILIRQVAAASHFRSTILHESKMLLKCQGVSDQAVAEALCSIMLLEESSPRQALTDFLLARKATIQKLLNQPHHGAGIKAQICSLVELLATTLKQAHALFYTLPEGLLPDPALPCGLLFSTLETITGQHPAGKGTGVLQEEMKLCSWFKHLPASIVEFQPTLRTLAHPISQEYLKDTLQKWIHMCNEDIKNGITNLLMYVKSMKGLAGIRDAMWELLTNESTNHSWDVLCRRLLEKPLLFWEDMMQQLFLDRLQTLTKEGFDSISSSSKELLVSALQELESSTSNSPSNKHIHFEYNMSLFLWSESPNDLPSDAAWVSVANRGQFASSGLSMKAQAISPCVQNFCSALDSKLKVKLDDLLAYLPSDDSSLPKDVSPTQAKSSAFDRYADAGTVQEMLRTQSVACIKHIVDCIRAELQSIEEGVQGQQDALNSAKLHSVLFMARLCQSLGELCPHLKQCILGKSESSEKPAREFRALRKQGKVKTQEIIPTQAKWQEVKEVLLQQSVMGYQVWSSAVVKVLIHGFTQSLLLDDAGSVLATATSWDELEIQEEAESGSSVTSKIRLPAQPSWYVQSFLFSLCQEINRVGGHALPKVTLQEMLKSCMVQVVAAYEKLSEEKQIKKEGAFPVTQNRALQLLYDLRYLNIVLTAKGDEVKSGRSKPDSRIEKVTDHLEALIDPFDLDVFTPHLNSNLHRLVQRTSVLFGLVTGTENQLAPRSSTFNSQEPHNILPLASSQIRFGLLPLSMTSTRKAKSTRNIETKAQVVPPARSTAGDPTVPGSLFRQLVSEEDNTSAPSLFKLGWLSSMTK
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9382
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.