missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cofilin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10960-1-AP-20UL
This item is not returnable.
View return policy
Description
Cofilin binds to F-actin and exhibits pH-sensitive F-actin depolymerizing activity. Regulates actin cytoskeleton dynamics. Important for normal progress through mitosis and normal cytokinesis. Plays a role in the regulation of cell morphology and cytoskeletal organization (By similarity). Required for the up-regulation of atypical chemokine receptor ACKR2 from endosomal compartment to cell membrane, increasing its efficiency in chemokine uptake and degradation. Required for neural tube morphogenesis and neural crest cell migration (By similarity).Specifications
| Cofilin | |
| Polyclonal | |
| Unconjugated | |
| CFL1 | |
| 18 kDa phosphoprotein, CFL, CFL1, Cofilin 1, cofilin 1 (non muscle), Cofilin, non muscle isoform, p18 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1072, 12631, 29271 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P18760, P23528, P45592 | |
| CFL1 | |
| Cofilin Fusion Protein Ag1399 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction