missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- No loss - reduced sample remains in the same crucible for analysis
- Immediate raw fiber extraction after fat extraction here
- Fatty components collected for easy disposal
- All reagents can be added through the same channel
Specifications
Specifications
| Product Type | Cold Extractor |
| Material | Stainless Steel |
| For Use With (Application) | Raw Fiber Analysis |
| Quantity | 1 Ea. |
| Height (Metric) | 300 mm |
| Width (Metric) | 730 mm |
| Weight (Metric) | 19 kg |
| Electrical Requirements | 230 V 50 Hz |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?

