missing translation for 'onlineSavingsMsg'
Learn More

Coenzyme Q10, TRC

Product Code. 30136244
Change view
Click to view available options
Quantity:
5 mg
100 mg
1 g
Unit Size:
100mg
1g
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136244 1 g 1g
30007739 100 mg 100mg
17907444 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136244 Supplier Toronto Research Chemicals Supplier No. TRCC6365011G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 303-98-0
Molecular Formula C59 H90 O4
Molecular Weight (g/mol) 863.34
Synonym Ubidecarenone, Ubiquinone, Q10, 2-[(all-E)-3,7,11,15,19,23,27,31,35,39-Decamethyltetraconta-2,6,10,14,18,22,26,30,34,38-decaenyl]-5,6-dimethoxy-3-methylbenzene-1,4-dione, 2,5-Cyclohexadiene-1,4-dione, 2-(3,7,11,15,19,23,27,31,35,39-decamethyl-2,6,10,14,18,22,26,30,34,38-tetracontadecaenyl)-5,6-dimethoxy-3-methyl-, (all-E)-, 2,5-Cyclohexadiene-1,4-dione, 2-[(2E,6E,10E,14E,18E,22E,26E,30E,34E)-3,7,11,15,19,23,27,31,35,39-decamethyl-2,6,10,14,18,22,26,30,34,38-tetracontadecaenyl]-5,6-dimethoxy-3-methyl- (9CI), Coenzyme Q10 (6CI), p-Benzoquinone, 2-(3,7,11,15,19,23,27,31,35,39-decamethyl-2,6,10,14,18,22,26,30,34,38-tetracontadecaenyl)-5,6-dimethoxy-3-methyl- (8CI), (all-E)-2-(3,7,11,15,19,23,27,31,35,39-Decamethyl-2,6,10,14,18,22,26,30,34,38-tetracontadecaenyl)-5,6-dimethoxy-3-methyl-2,5-cyclohexadiene-1,4-dione, Adelir, Aqua Q 10L10, Aqua Q10, Aqua-Q 10P40, Bio-Quinon, Bio-Quinone Q10, CoQ10, Cosmesome Q 10, Cudesan, Ensorb, Kaneka Q10, Kudesan, Li-Q-Sorb, Liquid-Q, NSC 140865, Neuquinon, Neuquinone, PureSorb Q 40, Q 10AA, Q-Gel, Q-Gel 100, Q-absorb, Ubidecarenone, Ubiquinone 10, Ubiquinone 50, Ubiquinone Q10, Unispheres Q 10, Vitamin Q
IUPAC Name 2-[(2E,6E,10E,14E,18E,22E,26E,30E,34E)-3,7,11,15,19,23,27,31,35,39-decamethyltetraconta-2,6,10,14,18,22,26,30,34,38-decaenyl]-5,6-dimethoxy-3-methylcyclohexa-2,5-diene-1,4-dione
SMILES COC1=C(OC)C(=O)C(=C(C)C1=O)C\C=C(/C)\CC\C=C(/C)\CC\C=C(/C)\CC\C=C(/C)\CC\C=C(/C)\CC\C=C(/C)\CC\C=C(/C)\CC\C=C(/C)\CC\C=C(/C)\CCC=C(C)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C59H90O4/c1-44(2)24-15-25-45(3)26-16-27-46(4)28-17-29-47(5)30-18-31-48(6)32-19-33-49(7)34-20-35-50(8)36-21-37-51(9)38-22-39-52(10)40-23-41-53(11)42-43-55-54(12)56(60)58(62-13)59(63-14)57(55)61/h24,26,28,30,32,34,36,38,40,42H,15-23,25,27,29,31,33,35,37,39,41,43H2,1-14H3/b45-26+,46-28+,47-30+,48-32+,49-34+,50-36+,51-38+,52-40+,53-42+
Formula Weight 862.6839
Percent Purity >95%
Chemical Name or Material Coenzyme Q10
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.