missing translation for 'onlineSavingsMsg'
Learn More

Coenzyme Q10, MedChemExpress

Product Code. 16472346
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
500 mg
1 g
5 g
Unit Size:
100mg
1g
200mg
500mg
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16472346 500 mg 500mg
16461486 200 mg 200mg
16442499 1 g 1g
16441308 5 g 5g
16438986 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16472346 Supplier Medchem Express Supplier No. HYN0111
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Coenzyme Q10 is an essential cofactor of the electron transport chain and a potent antioxidant agent.

Coenzyme Q10 is an essential cofactor of the electron transport chain and a potent antioxidant agent.

Specifications

Chemical Name or Material Coenzyme Q10
CAS 303-98-0
Color Orange
Purity Grade Notes Research
Molecular Formula C59H90O4
Quantity 500 mg
Synonym CoQ10 Ubiquinone-10
Solubility Information DMF : 40 mg/mL (46.33 mM; Need ultrasonic) ∣Ethanol : 2.5 mg/mL (2.90 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (1.16 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C1C(C/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)\C)=C(C)C(C(OC)=C1OC)=O
Molecular Weight (g/mol) 863.34
Formula Weight 863.34
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.